Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Syce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168244.1 | NP_001161716.1 | 4 | 1060 | ||
NM_001168246.1 | NP_001161718.1 | 5 | 759 | |||
NM_027954.3 | NP_082230.2 | 5 | 741 | |||
GenBank mRNA | AK005954.1 | - | 5 | 742 | ||
AK082833.1 | - | 4 | 627 | |||
AK167332.1 | - | 4 | 1059 | |||
BC094628.1 | - | 5 | 636 | 3374002 | ||
BC132534.1 | - | 5 | 691 | 40130811 |