Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Acad10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028037.4 | NP_082313.2 | 11 | 1643 | ||
GenBank mRNA | AK010568.1 | - | 1 | 94 | ||
AK050332.1 | - | 11 | 1847 | |||
AK148195.1 | - | 10 | 1452 | |||
BC027825.1 | - | 11 | 1619 | 5343012 | ||
BC029047.1 | - | 7 | 792 |