Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028044.2 | NP_082320.1 | 2 | 401 | ||
GenBank mRNA | AK005460.1 | - | 2 | 403 | ||
AK051628.1 | - | 2 | 419 | |||
AK081672.1 | - | 2 | 317 | |||
AK159555.1 | - | 2 | 446 | |||
AK159871.1 | - | 2 | 431 | |||
AK166578.1 | - | 2 | 409 | |||
AK167201.1 | - | 2 | 422 | |||
AK167613.1 | - | 2 | 419 | |||
AK169021.1 | - | 2 | 426 | |||
AK169098.1 | - | 2 | 446 | |||
AK169410.1 | - | 2 | 376 | |||
AK179383.1 | - | 2 | 132 | |||
AK180041.1 | - | 1 | 151 | |||
AK184231.1 | - | 2 | 137 | |||
AK208457.1 | - | 1 | 101 | |||
AK212516.1 | - | 1 | 50 | |||
AK216318.1 | - | 2 | 132 | |||
BC055711.1 | - | 2 | 402 | 6415164 | ||
BC085268.1 | - | 2 | 179 | 6487559 |