Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175229.3 | NP_780438.2 | 3, 4 | 486 | ||
GenBank mRNA | AK090123.1 | - | 3, 4 | 447 | ||
AK171001.1 | - | 4, 5 | 759 | |||
BC019803.1 | - | 4, 5 | 745 | |||
BC037186.1 | - | 4, 5 | 703 | |||
BC044058.1 | - | 3, 4 | 530 | |||
BC092355.1 | - | 4, 5 | 623 | |||
CJ178299.1 | - | 3, 4 | 454 |