Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ssna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023464.2 | NP_075953.1 | 2 | 284 | ||
GenBank mRNA | AB041656.1 | - | 2 | 278 | ||
AK088418.1 | - | 2 | 216 | |||
AK158412.1 | - | 401 | ||||
BC023033.1 | - | 2 | 247 | |||
BC037081.1 | - | 2 | 261 | 5098653 | ||
BC048360.1 | - | 2 | 262 | 5067090 | ||
BY703613.1 | - | 2 | 285 |