Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024209.2 | NP_077171.1 | 6 | 736 | ||
GenBank mRNA | AK002764.1 | - | 6 | 698 | ||
AK009104.1 | - | 6 | 716 | |||
BC002223.1 | - | 6 | 761 | 3491111 |