Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tsr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163718.1 | NP_001157190.1 | 4 | 607 | ||
NM_026676.3 | NP_080952.2 | 4 | 628 | |||
GenBank mRNA | AK002309.1 | - | 4 | 600 | ||
AK003675.1 | - | 2 | 1079 | |||
AK158743.1 | - | 4 | 629 | |||
AK169072.1 | - | 3 | 963 | |||
AK170074.1 | - | 4 | 597 | |||
BC027751.1 | - | 4 | 698 | 5011894 | ||
BC038884.1 | - | 2 | 401 | 1516275 | ||
BC089019.1 | - | 4 | 577 | 30551363 | ||
CA979987.1 | - | 4 | 556 |