Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SETMAR SIRNA Assays ›
          Gene Symbol:
          SETMAR
          Gene Name:
          SET domain and mariner transposase fusion gene
          Gene Aliases:
          METNASE, Mar1
          Chromosome Location:
          Chr.3: 4303304 - 4320649 on Build GRCh38
          UniGene ID:
          Hs.475300
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 18073

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SETMAR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243723.1 NP_001230652.1 2 481
          NM_001276325.1 NP_001263254.1 2 481
          NM_001320677.1 NP_001307606.1 2 481
          NM_006515.3 NP_006506.3 2 481
          XM_006713294.3 XP_006713357.1 2 543
          XM_017007032.1 XP_016862521.1 2 544
          GenBank mRNA AF054989.1 - 2 407
          AK222734.1 - 2 417
          AK297865.1 - 2 416
          AK302296.1 - 2 415
          AK307537.1 - 2 481
          AK311418.1 - 2 514
          AY952295.1 - 2 386
          BC011635.2 - 2 407 4080032
          BG479260.1 - 2 496
          DC357038.1 - 2 481
          U80776.1 - 2 398

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          viral or transposable element protein histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA double-strand break processing
          DNA catabolic process, endonucleolytic
          double-strand break repair via nonhomologous end joining
          cell proliferation
          histone H3-K36 methylation
          DNA integration
          replication fork processing
          mitotic DNA integrity checkpoint
          histone H3-K4 methylation
          negative regulation of cell cycle arrest
          nucleic acid phosphodiester bond hydrolysis
          histone H3-K36 dimethylation
          positive regulation of DNA topoisomerase (ATP-hydrolyzing) activity
          positive regulation of double-strand break repair via nonhomologous end joining
          negative regulation of chromosome organization
          single-stranded DNA endodeoxyribonuclease activity
          double-stranded DNA binding
          single-stranded DNA binding
          endonuclease activity
          protein binding
          zinc ion binding
          histone methyltransferase activity (H3-K4 specific)
          protein homodimerization activity
          structure-specific DNA binding
          DNA topoisomerase binding
          histone methyltransferase activity (H3-K36 specific)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-895sq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0