Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027117.3 | NP_081393.2 | 4 | 737 | ||
GenBank mRNA | AK009481.1 | - | 4 | 652 | ||
AK051418.1 | - | 4 | 664 | |||
AK164223.1 | - | 2 | 113 | |||
AY513272.1 | - | 4 | 688 | |||
BC005581.1 | - | 4 | 661 | 3968615 |