Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fyttd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159349.1 | NP_001152821.1 | 8 | 2786 | ||
NM_027226.4 | NP_081502.2 | 9 | 2888 | |||
GenBank mRNA | AK029785.1 | - | 9 | 2834 | ||
AK161252.1 | - | 9 | 2865 | |||
AK165633.1 | - | 9 | 2843 | |||
AK166032.1 | - | 9 | 2861 | |||
AK167564.1 | - | 1 | 1904 | |||
AK170511.1 | - | 9 | 2860 | |||
BC028253.1 | - | 9 | 2841 | 3983533 |