Search Thermo Fisher Scientific
Gene Symbol: Fam135b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177819.3 | NP_808487.2 | 20 | 4445 | ||
GenBank mRNA | AK005816.1 | - | 8 | 1313 | ||
BC119174.1 | - | 8 | 1061 | |||
BC137579.1 | - | 8 | 1061 |