Search
Search

Gltscr2| Gene Symbol: Gltscr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133831.3 | NP_598592.2 | 3 | 406 | ||
| GenBank mRNA | AK077341.1 | - | 3 | 360 | ||
| AK079167.1 | - | 3 | 360 | |||
| AK088461.1 | - | 3 | 363 | |||
| AK146916.1 | - | 3 | 375 | |||
| AK151836.1 | - | 3 | 363 | |||
| AK158665.1 | - | 3 | 306 | |||
| AK160070.1 | - | 3 | 362 | |||
| AK169290.1 | - | 3 | 366 | |||
| AY535001.1 | - | 3 | 351 | |||
| BC017637.1 | - | 3 | 382 | 4912415 | ||
| BC025810.1 | - | 3 | 313 | |||
| BY283382.1 | - | 3 | 406 | |||