Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006635.2 | NP_001006636.1 | 11 | 1152 | ||
NM_001319097.1 | NP_001306026.1 | 10 | 879 | |||
NM_001319098.1 | NP_001306027.1 | 9 | 871 | |||
NM_006554.4 | NP_006545.1 | 10 | 948 | |||
XM_011510497.2 | XP_011508799.1 | 11 | 1045 | |||
GenBank mRNA | AF053551.1 | - | 10 | 921 | ||
AK001287.1 | - | 10 | 900 | |||
AK289359.1 | - | 10 | 899 | |||
AK303564.1 | - | 11 | 1077 | |||
AK315918.1 | - | 11 | 1088 | |||
BC017271.2 | - | 11 | 1082 | 4635884 | ||
BC067831.1 | - | 10 | 927 | 5298149 | ||
BC088359.1 | - | 10 | 883 | 30378040 | ||
CX165048.1 | - | 10 | 772 |