Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rpl35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130484.1 | NP_001123956.1 | 3 | 188 | ||
NM_001130485.1 | NP_001123957.1 | 3 | 172 | |||
NM_021338.3 | NP_067313.2 | 3 | 218 | |||
GenBank mRNA | AK003063.1 | - | 3 | 142 | ||
AK010692.1 | - | 3 | 139 | |||
AK028214.1 | - | 3 | 142 | |||
AK046386.1 | - | 3 | 144 | |||
AK160963.1 | - | 3 | 144 | |||
AV486753.1 | - | 3 | 224 | |||
BB611247.1 | - | 3 | 140 | |||
BC027223.1 | - | 3 | 187 | 4036717 | ||
BC086771.1 | - | 2 | 127 | |||
BC090255.1 | - | 2 | 125 | 2655001 | ||
BC100332.1 | - | 109 | 30846392 | |||
BC100599.1 | - | 3 | 142 | 6762930 | ||
BU530423.1 | - | 3 | 179 | |||
CJ043213.1 | - | 3 | 189 | |||
Y16430.2 | - | 2 | 127 |
Gene Symbol: Ino80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC061495.1 | - | 4076 |