Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Stk25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021537.3 | NP_067512.3 | 3 | 311 | ||
GenBank mRNA | AF004934.1 | - | 2 | 149 | ||
AK148041.1 | - | 3 | 285 | |||
AK152917.1 | - | 3 | 254 | |||
BC029875.1 | - | 3 | 213 | |||
BC052913.1 | - | 3 | 246 | 30057101 | ||
BC071218.1 | - | 3 | 306 | 4012695 |