Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mrpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023167.2 | NP_075656.2 | 5 | 631 | ||
GenBank mRNA | AB041599.1 | - | 4 | 383 | ||
AB049634.1 | - | 5 | 630 | |||
AK002464.1 | - | 5 | 467 | |||
AK003746.2 | - | 5 | 588 | |||
AK216490.1 | - | 3 | 223 | |||
BC023078.1 | - | 5 | 412 | |||
BC037064.1 | - | 5 | 464 | |||
BC039983.1 | - | 5 | 599 | 4952757 | ||
BC051685.1 | - | 5 | 444 | |||
BC061095.1 | - | 5 | 447 | 6588836 |