Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Ostc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025509.3 | NP_079785.1 | 3 | 392 | ||
GenBank mRNA | AK009237.1 | - | 3 | 367 | ||
AK011085.1 | - | 3 | 344 | |||
AK013011.1 | - | 3 | 351 | |||
AK135332.1 | - | 3 | 348 | |||
BC014799.1 | - | 3 | 367 | |||
BC021935.1 | - | 3 | 366 | 5149713 |