Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trpd52l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025741.2 | NP_080017.1 | 1 | 911 | ||
GenBank mRNA | AK016362.1 | - | 1 | 900 | ||
AK016450.1 | - | 1 | 911 | |||
BC125314.1 | - | 1 | 911 | 40129829 | ||
BC125316.1 | - | 1 | 911 | 40129831 |