Search
Search

CSPG5| Gene Symbol: CSPG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206942.1 | NP_001193871.1 | 2 | 1111 | ||
| NM_001206943.1 | NP_001193872.1 | 2 | 1033 | |||
| NM_001206944.1 | NP_001193873.1 | 2 | 1033 | |||
| NM_001206945.1 | NP_001193874.1 | 2 | 1111 | |||
| NM_006574.3 | NP_006565.2 | 2 | 1033 | |||
| GenBank mRNA | AF059274.1 | - | 2 | 863 | ||
| AF461087.1 | - | 2 | 2956 | |||
| AF461088.1 | - | 1 | 965 | |||
| AF461089.1 | - | 2 | 2956 | |||
| AK294621.1 | - | 2 | 1111 | |||