Search
Search

Cryzl1| Gene Symbol: Cryzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026994.1 | NP_081270.1 | 12 | 1571 | ||
| NM_133679.2 | NP_598440.1 | 13 | 1616 | |||
| GenBank mRNA | AK008846.1 | - | 13 | 1389 | ||
| AK017995.1 | - | 12 | 1563 | |||
| AK145827.1 | - | 13 | 1440 | |||
| BC010479.1 | - | 13 | 1413 | 3968727 | ||
| BC019387.1 | - | 12 | 1380 | 4164847 | ||