Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sumf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145937.3 | NP_666049.2 | 9 | 2440 | ||
GenBank mRNA | AK151874.1 | - | 9 | 2436 | ||
AK159192.1 | - | 9 | 2429 | |||
AK161203.1 | - | 9 | 2433 | |||
AK207725.1 | - | 1 | 132 | |||
AK211675.1 | - | 1 | 132 | |||
AK216983.1 | - | 1 | 132 | |||
BC026981.1 | - | 9 | 2413 | 5365580 |