Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fbxw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164768.1 | NP_001158240.1 | 1 | 564 | ||
NM_001164769.1 | NP_001158241.1 | 3 | 623 | |||
NM_001164770.1 | NP_001158242.1 | 3 | 531 | |||
NM_001164772.1 | NP_001158244.1 | 3 | 623 | |||
NM_013890.4 | NP_038918.3 | 3 | 623 | |||
GenBank mRNA | AF140683.1 | - | 1 | 568 | ||
AK004015.1 | - | 3 | 589 | |||
AK005103.1 | - | 3 | 588 | |||
AK009893.1 | - | 2 | 645 | |||
AK033129.1 | - | 3 | 577 | |||
AK040764.1 | - | 3 | 590 | |||
AK078815.1 | - | 3 | 531 | |||
AK088898.1 | - | 1 | 303 | |||
AK133758.1 | - | 1 | 3591 | |||
AK145405.1 | - | 3 | 590 | |||
AK179719.1 | - | 1 | 117 | |||
AK184341.1 | - | 1 | 271 | |||
BC003834.1 | - | 3 | 709 | 3601733 | ||
X54352.1 | - | 2 | 553 |