Search Thermo Fisher Scientific
Gene Symbol: Schip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113420.1 | NP_001106891.1 | 5 | 733 | ||
NM_001113421.1 | NP_001106892.1 | 6 | 1417 | |||
NM_001282044.1 | NP_001268973.1 | 6 | 1400 | |||
NM_001282045.1 | NP_001268974.1 | 6 | 1258 | |||
NM_013928.5 | NP_038956.2 | 5 | 823 | |||
GenBank mRNA | AF145716.1 | - | 5 | 929 | ||
AK021361.1 | - | 5 | 684 | |||
AK083464.1 | - | 6 | 1243 | |||
AK146930.1 | - | 6 | 1374 | |||
AK168005.1 | - | 6 | 1327 | |||
BC060529.1 | - | 5 | 757 | |||
EU163407.1 | - | 6 | 1417 | |||
EU163408.1 | - | 5 | 733 | |||
KM233715.1 | - | 5 | 679 | |||
KM233716.1 | - | 5 | 684 |
Gene Symbol: Gm21949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113419.2 | NP_001106890.1 | 9 | 1595 | ||
GenBank mRNA | KJ941154.1 | - | 8 | 899 |