Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Grb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016719.1 | NP_057928.1 | 3 | 477 | ||
GenBank mRNA | AF155647.1 | - | 3 | 477 | ||
AK010849.1 | - | 3 | 494 | |||
AK010903.1 | - | 3 | 494 | |||
AK087758.1 | - | 2 | 228 | |||
AK147041.1 | - | 3 | 486 | |||
BC138581.1 | - | 3 | 383 | 8861601 | ||
BC145913.1 | - | 3 | 383 | 40131216 |