Search
Search

Nucb2| Gene Symbol: Nucb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130479.2 | NP_001123951.1 | 12 | 1350 | ||
| GenBank mRNA | AJ222586.1 | - | 12 | 1350 | ||
| AK145934.1 | - | 12 | 1305 | |||
| AK152672.1 | - | 6 | 587 | |||
| AK190340.1 | - | 3 | 182 | |||
| BC010459.1 | - | 12 | 1264 | 3585076 | ||