Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017401.2 | NP_059097.2 | 6 | 1045 | ||
GenBank mRNA | AF176098.1 | - | 6 | 818 | ||
AJ251804.2 | - | 6 | 789 | |||
BC056213.1 | - | 6 | 837 | 30024844 | ||
BC056996.1 | - | 3 | 2692 |