Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019727.2 | NP_062701.2 | 6 | 641 | ||
GenBank mRNA | AB019214.1 | - | 6 | 581 | ||
AF154120.1 | - | 6 | 645 | |||
AK149850.1 | - | 6 | 553 | |||
AK154079.1 | - | 6 | 554 | |||
AK155746.1 | - | 6 | 529 | |||
AK161241.1 | - | 6 | 622 | |||
AK167177.1 | - | 6 | 596 | |||
AK167993.1 | - | 6 | 536 | |||
AK168742.1 | - | 6 | 634 | |||
AK219007.1 | - | 4 | 171 | |||
BC066044.1 | - | 6 | 598 | 6807363 | ||
BC066189.1 | - | 6 | 532 | 30471747 |