Search
Search

Cpb2| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019775.3 | NP_062749.2 | 2, 3 | 216 | ||
| GenBank mRNA | AA048136.1 | - | 2, 3 | 216 | ||
| AB021968.1 | - | 2, 3 | 164 | |||
| AF164524.1 | - | 2, 3 | 231 | |||
| AF186188.1 | - | 2, 3 | 167 | |||
| AK004045.1 | - | 2, 3 | 189 | |||
| BC089577.1 | - | 2, 3 | 188 | 6766672 | ||