Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FBXW7 SIRNA Assays ›
          Gene Symbol:
          Fbxw7
          Gene Name:
          F-box and WD-40 domain protein 7
          Gene Aliases:
          1110001A17Rik, AGO, Cdc4, Fbw7, Fbwd6, Fbx30, Fbxo30, Fbxw6, SEL-10
          Chromosome Location:
          Chr.3: 84815577 - 84979198 on Build GRCm38
          UniGene ID:
          Mm.196475
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 186284

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fbxw7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177773.1 NP_001171244.1 11, 12 1809
          NM_001177774.1 NP_001171245.1 10, 11 1718
          NM_080428.3 NP_536353.2 8, 9 1354
          GenBank mRNA AB221593.1 - 8, 9 1196
          AF391192.1 - 8, 9 1354
          AF427101.1 - 8, 9 1276
          AK032174.1 - 2, 3 280
          BC060620.1 - 11, 12 1809
          BC066093.1 - 10, 11 1719
          BC085184.1 - 10, 11 3510 30664813
          BC131648.1 - 8, 9 1218 40110349

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          vasculogenesis
          regulation of transcription, DNA-templated
          cellular response to DNA damage stimulus
          sister chromatid cohesion
          Notch signaling pathway
          protein ubiquitination
          lung development
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of protein ubiquitination
          negative regulation of DNA endoreduplication
          cellular response to UV
          positive regulation of epidermal growth factor-activated receptor activity
          protein stabilization
          positive regulation of ubiquitin-protein transferase activity
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of proteasomal protein catabolic process
          regulation of mitophagy
          positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of protein targeting to mitochondrion
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          transcription factor activity, sequence-specific DNA binding
          protein binding
          cyclin binding
          protein binding, bridging
          ubiquitin protein ligase binding
          identical protein binding
          ubiquitin-protein transferase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bnpf6:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0