Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LILRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278426.3 | NP_001265355.2 | 7 | 990 | ||
NM_001278427.3 | NP_001265356.2 | 7 | 990 | |||
NM_001278428.3 | NP_001265357.2 | 7 | 990 | |||
NM_001278429.3 | NP_001265358.2 | 6 | 954 | |||
XM_017026215.1 | XP_016881704.1 | 8 | 1014 | |||
XM_017026216.1 | XP_016881705.1 | 7 | 987 | |||
XM_017026217.1 | XP_016881706.1 | 7 | 987 | |||
GenBank mRNA | AF025532.1 | - | 7 | 804 | ||
AF283988.1 | - | 7 | 814 | |||
AK292082.1 | - | 7 | 854 | |||
AK314783.1 | - | 6 | 803 | |||
BC026309.1 | - | 7 | 809 | 4795550 | ||
CR542048.1 | - | 7 | 775 | |||
U82979.1 | - | 7 | 775 | |||
U91925.1 | - | 7 | 820 |