Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TKT SIRNA Assays ›
          Gene Symbol:
          Tkt
          Gene Name:
          transketolase
          Gene Aliases:
          p68
          Chromosome Location:
          Chr.14: 30549131 - 30574724 on Build GRCm38
          UniGene ID:
          Mm.290692
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 186763

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tkt
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009388.6 NP_033414.1 4, 5 584
          GenBank mRNA AF195533.1 - 3, 4 250
          AK002627.1 - 4, 5 537
          AK012794.1 - 4, 5 539
          AK030446.1 - 4, 5 567
          AK140965.1 - 4, 5 568
          AK144146.1 - 4, 5 539
          AK145677.1 - 4, 5 537
          AK146157.1 - 4, 5 537
          AK150139.1 - 4, 5 537
          AK150769.1 - 4, 5 533
          AK150844.1 - 4, 5 533
          AK150856.1 - 4, 5 533
          AK150901.1 - 4, 5 537
          AK151916.1 - 4, 5 533
          AK151969.1 - 4, 5 533
          AK152245.1 - 4, 5 533
          AK152460.1 - 4, 5 553
          AK152886.1 - 4, 5 533
          AK159922.1 - 4, 5 537
          AK166763.1 - 4, 5 571
          AK167084.1 - 4, 5 571
          AK177246.1 - 3, 4 246
          AK181952.1 - 3, 4 350
          BC055336.1 - 4, 5 511 6413263
          U05809.1 - 4, 5 532

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_022592, XM_006252661, U09256.

          Panther Classification:

          Molecular Function -

          transketolase

          Gene Ontology Categories:

          Function(s) Process(es)

          pentose-phosphate shunt
          metabolic process
          pentose-phosphate shunt, non-oxidative branch
          regulation of growth
          glyceraldehyde-3-phosphate biosynthetic process
          ribose phosphate biosynthetic process
          magnesium ion binding
          catalytic activity
          transketolase activity
          transferase activity
          carbohydrate binding
          thiamine pyrophosphate binding
          protein homodimerization activity
          metal ion binding
          monosaccharide binding
          cofactor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fjwnj:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0