Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009437.4 | NP_033463.1 | 1, 2 | 616 | ||
GenBank mRNA | AK003953.1 | - | 1, 2 | 623 | ||
BC005644.1 | - | 1, 2 | 640 | 3257034 | ||
CT010210.1 | - | 1, 2 | 590 | |||
U35741.1 | - | 1, 2 | 604 |