Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Upk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009476.2 | NP_033502.2 | 2 | 197 | ||
GenBank mRNA | BC104260.1 | - | 2 | 197 | 40039813 | |
BC104261.1 | - | 2 | 197 | 40039814 | ||
CX565093.1 | - | 2 | 407 | |||
DQ358978.1 | - | 2 | 136 | |||
U08030.1 | - | 2 | 195 |