Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zfp37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290350.1 | NP_001277279.1 | 4 | 705 | ||
NM_001290351.1 | NP_001277280.1 | 4 | 696 | |||
NM_001290353.1 | NP_001277282.1 | 4 | 603 | |||
NM_009554.4 | NP_033580.3 | 4 | 693 | |||
GenBank mRNA | AK032469.1 | - | 4 | 575 | ||
BC024863.1 | - | 2 | 316 | |||
BC063757.1 | - | 4 | 577 | 6331110 | ||
CO430987.1 | - | 4 | 575 | |||
CO434318.1 | - | 4 | 530 | |||
X64413.1 | - | 4 | 499 | |||
X89264.1 | - | 5 | 605 |