Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TBX1 SIRNA Assays ›
          Gene Symbol:
          Tbx1
          Gene Name:
          T-box 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 18581704 - 18590671 on Build GRCm38
          UniGene ID:
          Mm.295194
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 187208

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tbx1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001285472.1 NP_001272401.1 6 845
          NM_001285476.1 NP_001272405.1 4 769
          NM_011532.2 NP_035662.1 4 773
          GenBank mRNA AF326960.2 - 4 773
          AF349658.1 - 6 845
          BC139474.1 - 4 773 8862496
          BC145636.1 - 4 769

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          Rel homology transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          blood vessel development
          patterning of blood vessels
          cell fate specification
          neural crest cell migration
          positive regulation of protein phosphorylation
          lymph vessel development
          blood vessel remodeling
          positive regulation of mesenchymal cell proliferation
          heart morphogenesis
          outflow tract septum morphogenesis
          outflow tract morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          multicellular organism development
          determination of left/right symmetry
          pattern specification process
          mesoderm development
          heart development
          muscle organ development
          sensory perception of sound
          cell proliferation
          positive regulation of cell proliferation
          anterior/posterior pattern specification
          vagus nerve morphogenesis
          epithelial cell differentiation
          thyroid gland development
          social behavior
          aorta morphogenesis
          ear morphogenesis
          inner ear morphogenesis
          outer ear morphogenesis
          middle ear morphogenesis
          odontogenesis of dentin-containing tooth
          muscle cell fate commitment
          positive regulation of MAPK cascade
          tongue morphogenesis
          cellular response to fibroblast growth factor stimulus
          negative regulation of cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          retinoic acid receptor signaling pathway
          blood vessel morphogenesis
          thymus development
          muscle organ morphogenesis
          embryonic cranial skeleton morphogenesis
          embryonic viscerocranium morphogenesis
          semicircular canal morphogenesis
          artery morphogenesis
          positive regulation of epithelial cell proliferation
          parathyroid gland development
          soft palate development
          pharyngeal system development
          face morphogenesis
          muscle tissue morphogenesis
          coronary artery morphogenesis
          enamel mineralization
          otic vesicle morphogenesis
          positive regulation of secondary heart field cardioblast proliferation
          cochlea morphogenesis
          mesenchymal cell apoptotic process
          regulation of organ morphogenesis
          positive regulation of tongue muscle cell differentiation
          negative regulation of mesenchymal cell apoptotic process
          RNA polymerase II intronic transcription regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein homodimerization activity
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-fj6pb:80/100.66.77.210:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0