Search
Search

ST3GAL2| Gene Symbol: ST3GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006927.3 | NP_008858.1 | 5, 6 | 1910 | ||
| GenBank mRNA | AK127322.1 | - | 4, 5 | 2875 | ||
| AK313020.1 | - | 4, 5 | 1041 | |||
| BC036777.1 | - | 4, 5 | 809 | 5750330 | ||
| U63090.1 | - | 4, 5 | 1447 | |||
| X96667.1 | - | 4, 5 | 1063 | |||