Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Polk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012048.2 | NP_036178.1 | 3 | 365 | ||
GenBank mRNA | AB027563.1 | - | 3 | 353 | ||
AF163571.1 | - | 3 | 365 | |||
AK042272.1 | - | 3 | 460 | |||
AK138299.1 | - | 3 | 368 | |||
AY769919.1 | - | 2 | 179 | |||
AY769920.1 | - | 2 | 179 | |||
AY769921.1 | - | 2 | 179 | |||
AY769922.1 | - | 2 | 179 | |||
AY769923.1 | - | 2 | 179 | |||
AY769924.1 | - | 2 | 179 | |||
AY769925.1 | - | 2 | 179 | |||
AY769926.1 | - | 2 | 179 | |||
AY769927.1 | - | 2 | 179 | |||
AY769928.1 | - | 2 | 179 |