Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyro3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290800.1 | NP_001277729.1 | 9 | 1882 | ||
NM_019392.2 | NP_062265.2 | 9 | 1418 | |||
GenBank mRNA | AB000828.1 | - | 9 | 1155 | ||
AK134607.1 | - | 1 | 255 | |||
AK141198.1 | - | 9 | 1189 | |||
BC066058.1 | - | 9 | 1418 | 6834015 | ||
BC082325.1 | - | 9 | 1366 | 30356080 | ||
D17393.1 | - | 9 | 1860 | |||
U05683.1 | - | 9 | 1203 | |||
U18342.1 | - | 9 | 1335 | |||
U18343.1 | - | 9 | 1878 | |||
U18933.1 | - | 9 | 1354 | |||
X78103.1 | - | 9 | 1362 |