Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021885.4 | NP_068685.1 | 9, 10 | 1331 | ||
GenBank mRNA | AK147700.1 | - | 9, 10 | 1386 | ||
BC096554.1 | - | 9, 10 | 1312 | 30616697 | ||
U52433.1 | - | 9, 10 | 1319 | |||
U52824.1 | - | 10, 11 | 1464 | |||
U54643.1 | - | 9, 10 | 1249 |