Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trim24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272076.1 | NP_001259005.1 | 9 | 1301 | ||
NM_145076.4 | NP_659542.3 | 9 | 1721 | |||
GenBank mRNA | AB221637.1 | - | 9 | 1421 | ||
AK084870.1 | - | 8 | 1593 | |||
AK133144.1 | - | 9 | 1301 | |||
AK166538.1 | - | 5 | 678 | |||
BC056959.1 | - | 9 | 1699 | 5698079 | ||
S78221.1 | - | 9 | 1721 |