Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006851.2 | NP_006842.2 | 2, 3 | 576 | ||
XM_017018721.1 | XP_016874210.1 | 2, 3 | 456 | |||
GenBank mRNA | AF400440.1 | - | 2, 3 | 456 | ||
AK313449.1 | - | 2, 3 | 442 | |||
BC012510.1 | - | 2, 3 | 492 | 4453841 | ||
U16307.3 | - | 2, 3 | 512 | |||
X91911.1 | - | 2, 3 | 556 |