Search
Search

Slc1a6| Gene Symbol: Slc1a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032065.1 | NP_114454.1 | 5 | 760 | ||
| XM_008765182.1 | XP_008763404.1 | 5 | 934 | |||
| GenBank mRNA | AY286330.1 | - | 5 | 754 | ||
| BC098912.1 | - | 5 | 784 | 7315936 | ||
| U80915.1 | - | 3 | 390 | |||
| U89608.1 | - | 5 | 760 | |||