Search
Search

Bnip3| Gene Symbol: Bnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053420.3 | NP_445872.1 | 3 | 346 | ||
| GenBank mRNA | AF243515.1 | - | 3 | 310 | ||
| AY095482.1 | - | 3 | 310 | |||
| BC070958.1 | - | 3 | 304 | 7105707 | ||
| BC081690.1 | - | 3 | 284 | 7190121 | ||
| CO403082.1 | - | 3 | 382 | |||
| FQ213448.1 | - | 2 | 107 | |||
| FQ215108.1 | - | 3 | 316 | |||
| FQ215127.1 | - | 3 | 316 | |||
| FQ215203.1 | - | 3 | 337 | |||
| FQ216745.1 | - | 3 | 337 | |||
| FQ216807.1 | - | 3 | 337 | |||
| FQ220853.1 | - | 3 | 315 | |||
| FQ221849.1 | - | 3 | 337 | |||