Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NTRK1 SIRNA Assays ›
Gene Symbol:
Ntrk1
Gene Name:
neurotrophic tyrosine kinase, receptor, type 1
Gene Aliases:
Trk
Chromosome Location:
Chr.2: 187143562 - 187160373 on Build Rnor_6.0
UniGene ID:
Rn.39098
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
191893
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ntrk1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021589.1
NP_067600.1
1, 2
270
XM_006232746.2
XP_006232808.1
1, 2
261
GenBank mRNA
M85214.1
-
1, 2
270
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
nervous system development
axon guidance
aging
learning or memory
circadian rhythm
negative regulation of cell proliferation
response to radiation
programmed cell death involved in cell development
positive regulation of neuron projection development
response to activity
peptidyl-tyrosine phosphorylation
sensory perception of pain
olfactory nerve development
B cell differentiation
response to nutrient levels
response to nicotine
nerve growth factor signaling pathway
mechanoreceptor differentiation
response to drug
positive regulation of programmed cell death
negative regulation of neuron apoptotic process
response to ethanol
positive regulation of angiogenesis
positive regulation of Ras protein signal transduction
protein autophosphorylation
neurotrophin TRK receptor signaling pathway
sympathetic nervous system development
response to axon injury
detection of temperature stimulus involved in sensory perception of pain
detection of mechanical stimulus involved in sensory perception of pain
positive regulation of NF-kappaB transcription factor activity
response to hydrostatic pressure
response to electrical stimulus
positive regulation of synaptic transmission, glutamatergic
Sertoli cell development
axonogenesis involved in innervation
positive regulation of ERK1 and ERK2 cascade
cellular response to nicotine
cellular response to growth factor stimulus
negative regulation of neuron death
cellular response to nerve growth factor stimulus
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
neurotrophin p75 receptor binding
protein binding
ATP binding
nerve growth factor receptor activity
kinase binding
protein homodimerization activity
ephrin receptor binding
nerve growth factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0