Search
Search

Dap| Gene Symbol: Dap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022526.2 | NP_071971.1 | 2 | 332 | ||
| GenBank mRNA | BC060569.1 | - | 2 | 150 | 6890457 | |
| FQ218203.1 | - | 2 | 217 | |||
| FQ228698.1 | - | 2 | 170 | |||
| FQ228865.1 | - | 2 | 176 | |||
| U05334.1 | - | 2 | 333 | |||