Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008396.2 | NP_001008397.1 | 25 | 3704 | ||
NM_007086.3 | NP_009017.1 | 26 | 3797 | |||
XM_006720012.2 | XP_006720075.1 | 26 | 3789 | |||
XM_011536373.2 | XP_011534675.1 | 25 | 3596 | |||
XM_017020940.1 | XP_016876429.1 | 25 | 3702 | |||
XM_017020941.1 | XP_016876430.1 | 23 | 3400 | |||
GenBank mRNA | AJ006266.1 | - | 25 | 3745 | ||
AK001585.1 | - | 16 | 2720 | |||
AK001620.1 | - | 9 | 1524 | |||
AY517556.1 | - | 1 | 247 |