Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cxcl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145672.4 | NP_663705.1 | 3 | 306 | ||
GenBank mRNA | AF462610.1 | - | 3 | 213 | ||
AF537208.1 | - | 3 | 306 | |||
BC087594.1 | - | 255 | 7310193 |