Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PYCARD SIRNA Assays ›
Gene Symbol:
Pycard
Gene Name:
PYD and CARD domain containing
Gene Aliases:
Asc
Chromosome Location:
Chr.1: 199438029 - 199439062 on Build Rnor_6.0
UniGene ID:
Rn.7817
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
194186
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pycard
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_172322.1
NP_758825.1
3
485
GenBank mRNA
AB053165.1
-
3
485
FQ226280.1
-
3
643
FQ227544.1
-
3
665
Back To Top
More Information
Panther Classification:
Molecular Function -
protease
Gene Ontology Categories:
Function(s)
Process(es)
myeloid dendritic cell activation
activation of innate immune response
positive regulation of defense response to virus by host
myeloid dendritic cell activation involved in immune response
positive regulation of antigen processing and presentation of peptide antigen via MHC class II
positive regulation of adaptive immune response
activation of cysteine-type endopeptidase activity involved in apoptotic process
inflammatory response
response to bacterium
regulation of autophagy
regulation of tumor necrosis factor-mediated signaling pathway
positive regulation of actin filament polymerization
regulation of protein stability
negative regulation of NF-kappaB transcription factor activity
interleukin-1 beta production
negative regulation of interferon-beta production
positive regulation of interferon-gamma production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
tumor necrosis factor-mediated signaling pathway
positive regulation of activated T cell proliferation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
macropinocytosis
positive regulation of JNK cascade
positive regulation of interleukin-1 beta secretion
regulation of inflammatory response
positive regulation of phagocytosis
defense response to Gram-negative bacterium
positive regulation of T cell activation
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
defense response to virus
positive regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to tumor necrosis factor
negative regulation of protein serine/threonine kinase activity
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of chemokine secretion
positive regulation of release of cytochrome c from mitochondria
positive regulation of T cell migration
positive regulation of interleukin-8 secretion
positive regulation of interleukin-6 secretion
positive regulation of cysteine-type endopeptidase activity
positive regulation of interleukin-10 secretion
positive regulation of extrinsic apoptotic signaling pathway
regulation of intrinsic apoptotic signaling pathway
protease binding
protein binding
Pyrin domain binding
protein homodimerization activity
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-b8kr6:80/100.66.77.153:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline