Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIF SIRNA Assays ›
          Gene Symbol:
          Lif
          Gene Name:
          leukemia inhibitory factor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 84492909 - 84500550 on Build Rnor_6.0
          UniGene ID:
          Rn.44379
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 194515

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lif
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022196.2 NP_071532.2 2 146
          XM_006251364.2 XP_006251426.1 3 281
          XM_008770359.1 XP_008768581.1 2 851
          GenBank mRNA AB010275.1 - 2 146
          M32748.1 - 2 146

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel remodeling
          transcription from RNA polymerase II promoter
          immune response
          embryo implantation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of gene expression
          positive regulation of neuron projection development
          negative regulation of angiogenesis
          stem cell population maintenance
          lung development
          organ regeneration
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          tyrosine phosphorylation of Stat3 protein
          positive regulation of tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of MAPK cascade
          regulation of cell differentiation
          positive regulation of macrophage differentiation
          negative regulation of meiotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          decidualization
          negative regulation of hormone secretion
          lung alveolus development
          muscle organ morphogenesis
          neuron development
          astrocyte differentiation
          positive regulation of astrocyte differentiation
          leukemia inhibitory factor signaling pathway
          stem cell differentiation
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of corticotropin secretion
          retina development in camera-type eye
          maternal process involved in female pregnancy
          transdifferentiation
          lung vasculature development
          lung lobe morphogenesis
          trophoblast giant cell differentiation
          spongiotrophoblast differentiation
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          regulation of metanephric nephron tubule epithelial cell differentiation
          positive regulation of protein localization to nucleus
          positive regulation of histone H3-K27 acetylation
          regulation of RNA polymerase II regulatory region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II transcription factor recruiting
          receptor binding
          cytokine activity
          leukemia inhibitory factor receptor binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0