Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTGF SIRNA Assays ›
Gene Symbol:
Ctgf
Gene Name:
connective tissue growth factor
Gene Aliases:
CTGRP
Chromosome Location:
Chr.1: 21851657 - 21854773 on Build Rnor_6.0
UniGene ID:
Rn.17145
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
194555
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ctgf
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022266.2
NP_071602.1
5
1361
GenBank mRNA
AF120275.1
-
5
1361
BC072503.1
-
4
1730
7098774
FQ230151.1
-
1
15
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
cartilage condensation
ossification
angiogenesis
regulation of cell growth
tissue homeostasis
positive regulation of protein phosphorylation
cell adhesion
cell-matrix adhesion
integrin-mediated signaling pathway
cell-cell signaling
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
response to glucose
organ senescence
positive regulation of gene expression
negative regulation of gene expression
positive regulation of cell death
response to organic cyclic compound
cell migration
cell differentiation
lung development
regulation of chondrocyte differentiation
response to estradiol
positive regulation of collagen biosynthetic process
response to anoxia
intracellular signal transduction
chondrocyte proliferation
response to amino acid
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to peptide hormone
positive regulation of cell differentiation
positive regulation of JNK cascade
positive regulation of cell activation
response to mineralocorticoid
positive regulation of stress fiber assembly
cytosolic calcium ion transport
positive regulation of cardiac muscle contraction
negative regulation of cell death
connective tissue development
extracellular matrix constituent secretion
positive regulation of G0 to G1 transition
positive regulation of ERK1 and ERK2 cascade
response to fatty acid
DNA biosynthetic process
reactive oxygen species metabolic process
fibronectin binding
integrin binding
insulin-like growth factor binding
growth factor activity
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-7d94m:80/100.66.78.9:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0